Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271705.1 | NP_001258634.1 | 3 | 217 | ||
NM_008827.3 | NP_032853.1 | 3 | 550 | |||
GenBank mRNA | AK042891.1 | - | 3 | 534 | ||
AK088943.1 | - | 3 | 533 | |||
BC016567.1 | - | 3 | 522 | 2649577 | ||
X80171.1 | - | 3 | 318 | |||
X96793.1 | - | 3 | 517 |