Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008839.2 | NP_032865.2 | 1 | 224 | ||
GenBank mRNA | BC089038.1 | - | 1 | 224 | 30692343 | |
BC130228.1 | - | 1 | 173 | 40141870 | ||
U03279.1 | - | 1 | 169 |