Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008856.3 | NP_032882.2 | 2, 3 | 542 | ||
GenBank mRNA | AK076352.1 | - | 2, 3 | 741 | ||
AK145817.1 | - | 2, 3 | 788 | |||
AK155951.1 | - | 3, 4 | 885 | |||
AK164044.1 | - | 2, 3 | 741 | |||
BC031121.1 | - | 2, 3 | 542 | 5059633 | ||
D90242.1 | - | 2, 3 | 479 |