Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008866.2 | NP_032892.1 | 5 | 278 | ||
GenBank mRNA | AK002674.1 | - | 5 | 282 | ||
AK050549.1 | - | 5 | 255 | |||
AK146874.1 | - | 5 | 293 | |||
AK167231.1 | - | 300 | ||||
BC013536.1 | - | 5 | 278 | 4240573 | ||
BC052848.1 | - | 5 | 319 | 30055025 | ||
CT010201.1 | - | 5 | 257 | |||
U89352.1 | - | 5 | 257 |