Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008856.3 | NP_032882.2 | 3 | 609 | ||
GenBank mRNA | AK076352.1 | - | 3 | 808 | ||
AK145817.1 | - | 3 | 855 | |||
AK155951.1 | - | 4 | 952 | |||
AK164044.1 | - | 3 | 808 | |||
BC031121.1 | - | 3 | 609 | 5059633 | ||
D90242.1 | - | 3 | 546 |