Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030882.3 | NP_112092.2 | 5 | 1767 | ||
NM_145637.2 | NP_663612.2 | 6 | 1908 | |||
XM_011530074.1 | XP_011528376.1 | 6 | 2474 | |||
XM_011530075.1 | XP_011528377.1 | 7 | 2480 | |||
XM_011530076.2 | XP_011528378.1 | 6 | 1785 | |||
XM_011530077.2 | XP_011528379.1 | 5 | 1863 | |||
XM_011530078.1 | XP_011528380.1 | 7 | 2574 | |||
XM_017028724.1 | XP_016884213.1 | 6 | 1782 | |||
GenBank mRNA | AB208990.1 | - | 5 | 1662 | ||
AF305225.1 | - | 5 | 1651 | |||
AF305429.1 | - | 5 | 1625 | |||
AF324223.1 | - | 5 | 1767 | |||
AF324224.1 | - | 6 | 1908 | |||
AK056938.1 | - | 5 | 1696 | |||
BC004395.1 | - | 5 | 1634 | 3636011 |