Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008810.2 | NP_032836.1 | 11 | 1712 | ||
GenBank mRNA | AK145718.1 | - | 11 | 1790 | ||
AK148459.1 | - | 11 | 1733 | |||
AK168152.1 | - | 11 | 1763 | |||
AK169289.1 | - | 11 | 1787 | |||
BC007142.1 | - | 11 | 1712 | 3600605 |