Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008881.2 | NP_032907.1 | 16 | 3399 | ||
GenBank mRNA | AK220460.1 | - | 13 | 1639 | ||
BC138023.1 | - | 15 | 3191 | 8861042 | ||
D86948.1 | - | 16 | 3383 |