Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008883.2 | NP_032909.2 | 31 | 5525 | ||
GenBank mRNA | AK049319.1 | - | 13 | 1939 | ||
AK220285.1 | - | 27 | 4149 | |||
BC093482.1 | - | 31 | 5277 | 30945372 | ||
BC127155.1 | - | 1 | 416 | |||
D86950.1 | - | 31 | 5491 |