Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163557.1 | NP_001157029.1 | 3 | 392 | ||
NM_008905.2 | NP_032931.2 | 3 | 628 | |||
GenBank mRNA | AK035364.1 | - | 3 | 396 | ||
AK135964.1 | - | 3 | 663 | |||
AK171349.1 | - | 3 | 473 | |||
BB627669.1 | - | 3 | 393 | |||
BC051202.1 | - | 3 | 476 | |||
BC086931.1 | - | 3 | 747 | |||
BC100346.1 | - | 3 | 392 | 30936010 | ||
U79024.1 | - | 3 | 413 |