Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015002.1 | - | 2 | 317 | ||
AK133393.1 | - | 2 | 338 | |||
BC046617.1 | - | 2 | 143 | |||
BC141079.1 | - | 2 | 158 | 9055691 | ||
BC145347.1 | - | 2 | 158 | |||
BY716864.1 | - | 2 | 364 | |||
M81475.1 | - | 2 | 311 |