Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008891.2 | NP_032917.2 | 2, 3 | 277 | ||
GenBank mRNA | AK009055.1 | - | 2, 3 | 232 | ||
AK028219.1 | - | 2, 3 | 218 | |||
AK030978.1 | - | 2, 3 | 206 | |||
AK050669.1 | - | 2, 3 | 223 | |||
AK051640.1 | - | 2, 3 | 275 | |||
AK085295.1 | - | 2, 3 | 234 | |||
AK157587.1 | - | 2, 3 | 221 | |||
AK160616.1 | - | 2, 3 | 212 | |||
AK161109.1 | - | 2, 3 | 217 | |||
BC132571.1 | - | 2, 3 | 211 | 40130848 | ||
BC138014.1 | - | 2, 3 | 211 | 8861033 | ||
BY775218.1 | - | 2, 3 | 278 | |||
Y08701.1 | - | 2, 3 | 211 |