Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320559.1 | NP_001307488.1 | 5 | 3102 | ||
NM_001320560.1 | NP_001307489.1 | 5 | 2903 | |||
NM_033027.3 | NP_149016.2 | 5 | 2971 | |||
XM_017007048.1 | XP_016862537.1 | 5 | 6067 | |||
XM_017007049.1 | XP_016862538.1 | 3 | 2355 | |||
GenBank mRNA | AB053121.1 | - | 5 | 2948 | ||
AL117565.1 | - | 1 | 1174 | |||
BC028213.1 | - | 3 | 1731 | |||
BC038949.1 | - | 5 | 2918 | 5547938 |