Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033050.5 | NP_149039.2 | 3 | 1303 | ||
GenBank mRNA | AF247785.1 | - | 4 | 1187 | ||
AF348078.1 | - | 2 | 1240 | |||
AK290420.1 | - | 3 | 1303 | |||
BC030948.1 | - | 3 | 1297 | 4594810 |
Set Membership: |
G-protein coupled receptor |