Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145811.1 | NP_001139283.1 | 15 | 2512 | ||
NM_001145819.1 | NP_001139291.1 | 16 | 2474 | |||
NM_017508.2 | NP_059978.1 | 15 | 2534 | |||
NM_033326.3 | NP_201583.2 | 16 | 2420 | |||
GenBank mRNA | AF309034.1 | - | 16 | 2403 | ||
AL136780.1 | - | 15 | 2513 | |||
BC047064.1 | - | 16 | 2470 | 5269193 |