Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090006.1 | - | 4, 5 | 331 | ||
AK184128.1 | - | 3, 4 | 305 | |||
BC051387.1 | - | 4, 5 | 374 | 4460290 | ||
S63463.1 | - | 4, 5 | 446 | |||
X17459.1 | - | 4, 5 | 441 | |||
Y18248.1 | - | 1, 2 | 58 |