Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009052.2 | NP_033078.2 | 3 | 716 | ||
GenBank mRNA | AF051347.1 | - | 3 | 649 | ||
AF097438.1 | - | 3 | 661 | |||
AK003429.1 | - | 1 | 331 | |||
BC058805.1 | - | 1 | 988 | |||
BC061179.1 | - | 3 | 661 | 6531808 | ||
BC089464.1 | - | 3 | 710 | 6594252 | ||
CB194115.1 | - | 3 | 638 |