Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009055.4 | NP_033081.3 | 13 | 1956 | ||
GenBank mRNA | AK037226.1 | - | 6 | 899 | ||
AK039651.1 | - | 1 | 340 | |||
AK083571.1 | - | 13 | 1927 | |||
AK147790.1 | - | 13 | 1961 | |||
BC057018.1 | - | 13 | 1935 | 5705637 | ||
X76088.1 | - | 13 | 1863 |