Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159725.2 | NP_001153197.2 | 3, 4 | 480 | ||
NM_008998.4 | NP_033024.1 | 3, 4 | 711 | |||
GenBank mRNA | AB232611.1 | - | 2, 3 | 317 | ||
AK009707.1 | - | 3, 4 | 481 | |||
AK075591.1 | - | 3, 4 | 712 | |||
BC013170.1 | - | 3, 4 | 560 | 2648145 | ||
BC051071.1 | - | 3, 4 | 687 | 6307319 | ||
X70804.1 | - | 2, 3 | 317 |