Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009076.3 | NP_033102.2 | 4 | 374 | ||
GenBank mRNA | AK002973.1 | - | 4 | 373 | ||
AK008347.1 | - | 4 | 373 | |||
AK012349.1 | - | 4 | 373 | |||
AK012428.1 | - | 4 | 371 | |||
AK088464.1 | - | 4 | 373 | |||
AK144905.1 | - | 4 | 373 | |||
AK150318.1 | - | 4 | 374 | |||
AK151532.1 | - | 4 | 373 | |||
AK151592.1 | - | 4 | 373 | |||
AK167761.1 | - | 4 | 393 | |||
AK168121.1 | - | 4 | 373 | |||
AK194205.1 | - | 2 | 134 | |||
AK198056.1 | - | 2 | 134 | |||
BC018321.1 | - | 4 | 370 | 3583073 | ||
BC081469.1 | - | 4 | 357 | 6824411 | ||
BC090393.1 | - | 352 | 5011867 | |||
BY295120.1 | - | 4 | 376 | |||
L04280.1 | - | 4 | 361 |