Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009085.2 | NP_033111.2 | 4 | 396 | ||
GenBank mRNA | AK132114.1 | - | 4 | 338 | ||
AK146240.1 | - | 4 | 346 | |||
BC130018.1 | - | 4 | 397 | 40140532 |