Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009031.3 | NP_033057.3 | 12 | 1598 | ||
GenBank mRNA | AK076016.1 | - | 12 | 1523 | ||
AK078349.1 | - | 12 | 1511 | |||
AK082892.1 | - | 12 | 1510 | |||
AK135779.1 | - | 12 | 1512 | |||
AK135869.1 | - | 11 | 1207 | |||
AK135875.1 | - | 11 | 1207 | |||
AK135956.1 | - | 12 | 1512 | |||
AK136110.1 | - | 12 | 1510 | |||
AK145531.1 | - | 12 | 1509 | |||
AK145651.1 | - | 12 | 1492 | |||
AK146014.1 | - | 12 | 1510 | |||
AK146439.1 | - | 12 | 1569 | |||
AK146904.1 | - | 12 | 1511 | |||
AK146967.1 | - | 12 | 1510 | |||
AK147062.1 | - | 12 | 1569 | |||
AK148852.1 | - | 12 | 1538 | |||
AK153913.1 | - | 12 | 1555 | |||
AK160023.1 | - | 12 | 1510 | |||
AK181761.1 | - | 1 | 52 | |||
AK213484.1 | - | 2 | 185 | |||
AK214411.1 | - | 4 | 261 | |||
AK214866.1 | - | 4 | 259 | |||
AK215897.1 | - | 4 | 261 | |||
BC003785.1 | - | 12 | 1527 | 3600013 | ||
U35142.1 | - | 12 | 1537 |