Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selplg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009151.3 | NP_033177.3 | 2 | 316 | ||
GenBank mRNA | AK089188.1 | - | 2 | 291 | ||
AK089214.1 | - | 2 | 242 | |||
AK155762.1 | - | 2 | 247 | |||
AK156933.1 | - | 2 | 247 | |||
AK158134.1 | - | 2 | 247 | |||
AK171693.1 | - | 2 | 260 | |||
AK172077.1 | - | 2 | 250 | |||
BC003874.1 | - | 2 | 248 | 3495181 | ||
CJ134610.1 | - | 2 | 317 | |||
X91144.1 | - | 1 | 165 |