Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009186.4 | NP_033212.1 | 2 | 268 | ||
GenBank mRNA | AK049573.1 | - | 2 | 269 | ||
AK150007.1 | - | 2 | 269 | |||
AK152519.1 | - | 2 | 263 | |||
AK152761.1 | - | 2 | 269 | |||
AK167387.1 | - | 2 | 267 | |||
AK167544.1 | - | 2 | 269 | |||
BC057996.1 | - | 3 | 444 | |||
BC061177.1 | - | 2 | 265 | 6594226 | ||
BC086795.1 | - | 2 | 255 | 6828358 | ||
X80232.1 | - | 2 | 260 |