Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009129.2 | NP_033155.1 | 2 | 925 | ||
GenBank mRNA | AK002953.1 | - | 1 | 422 | ||
BC014717.1 | - | 2 | 894 | |||
BC048249.1 | - | 2 | 825 | 6493827 |