Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009139.3 | NP_033165.1 | 4 | 493 | ||
GenBank mRNA | AF128188.1 | - | 4 | 389 | ||
AF128189.1 | - | 4 | 389 | |||
AF128190.1 | - | 4 | 389 | |||
AF128191.1 | - | 4 | 389 | |||
AF128192.1 | - | 4 | 389 | |||
AK002697.1 | - | 4 | 396 | |||
AK008547.1 | - | 4 | 397 | |||
AK150336.1 | - | 4 | 396 | |||
AK154553.1 | - | 4 | 397 | |||
BC002073.1 | - | 4 | 386 | 3492808 | ||
M58004.1 | - | 4 | 396 |