Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113478.1 | NP_001106950.1 | 16, 17 | 1872 | ||
NM_009146.3 | NP_033172.2 | 16, 17 | 1834 | |||
GenBank mRNA | AK075573.1 | - | 16, 17 | 1825 | ||
AK144933.1 | - | 16, 17 | 1826 | |||
AK162693.1 | - | 16, 17 | 1827 | |||
AK167289.1 | - | 16, 17 | 1862 | |||
BC027770.1 | - | 15, 16 | 1832 | 5344922 | ||
D50464.1 | - | 16, 17 | 1822 |