Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009210.3 | NP_033236.2 | 2 | 378 | ||
GenBank mRNA | AB221636.1 | - | 2 | 201 | ||
AF010600.1 | - | 2 | 378 | |||
AF165911.1 | - | 2 | 357 | |||
AK141235.1 | - | 2 | 371 | |||
AK142961.1 | - | 2 | 335 | |||
AK149897.1 | - | 2 | 416 | |||
AK166911.1 | - | 2 | 331 | |||
BC057116.1 | - | 2 | 347 | 6408672 | ||
BC059240.1 | - | 2 | 347 | 6408672 | ||
BY007037.1 | - | 2 | 385 | |||
BY726838.1 | - | 2 | 336 |