Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009247.2 | NP_033273.1 | 2 | 620 | ||
GenBank mRNA | AK004999.1 | - | 2 | 622 | ||
AK146576.1 | - | 822 | ||||
BC061176.1 | - | 2 | 631 | 30381238 | ||
M75717.1 | - | 2 | 615 |