siRNA Details
Gene Symbol: Sox11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009234.6 | NP_033260.4 | 1 | 1371 | ||
GenBank mRNA | AB108675.1 | - | 1 | 1061 | ||
AF009414.1 | - | 1 | 1349 | |||
AK146514.1 | - | 1 | 1331 | |||
AK165005.1 | - | 1 | 1371 | |||
AK165693.1 | - | 1 | 1366 | |||
BC053503.1 | - | 1 | 1227 | |||
BC062095.1 | - | 1 | 1174 | 5716171 | ||
BC078643.1 | - | 1 | 1356 | 30144974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
