Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009287.4 | NP_033313.2 | 6 | 1313 | ||
GenBank mRNA | AK041944.1 | - | 6 | 1332 | ||
AK194819.1 | - | 260 | ||||
AK200235.1 | - | 3 | 260 | |||
BC021644.1 | - | 6 | 1315 | 4191156 | ||
U47323.1 | - | 6 | 915 |