Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp354a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009329.3 | NP_033355.2 | 5 | 592 | ||
GenBank mRNA | AB221586.1 | - | 4 | 396 | ||
AF184111.1 | - | 5 | 554 | |||
AK030469.1 | - | 5 | 571 | |||
BC050843.1 | - | 5 | 592 | 6506256 | ||
BC087540.1 | - | 5 | 728 | 4239789 | ||
L77247.1 | - | 4 | 456 |