Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009360.4 | NP_033386.1 | 5 | 625 | ||
GenBank mRNA | AK017974.1 | - | 5 | 653 | ||
AK050446.1 | - | 5 | 626 | |||
AK161747.1 | - | 5 | 652 | |||
AK163563.1 | - | 5 | 643 | |||
AK167348.1 | - | 5 | 626 | |||
AK167777.1 | - | 5 | 652 | |||
AK169808.1 | - | 5 | 653 | |||
AK178048.1 | - | 2 | 125 | |||
AK187847.1 | - | 2 | 226 | |||
AK213712.1 | - | 2 | 200 | |||
BC001987.1 | - | 5 | 620 | 3487433 | ||
BC083084.1 | - | 5 | 620 | 6514962 | ||
L07107.1 | - | 4 | 422 | |||
U57939.1 | - | 5 | 617 | |||
U63858.1 | - | 5 | 459 | |||
U63859.1 | - | 5 | 825 | |||
U63860.1 | - | 3 | 253 |