Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289468.1 | NP_001276397.1 | 3 | 1072 | ||
NM_009337.3 | NP_033363.1 | 4 | 1142 | |||
GenBank mRNA | AF031956.1 | - | 4 | 1119 | ||
AK149361.1 | - | 3 | 1054 | |||
AK163387.1 | - | 4 | 1126 | |||
BC052336.1 | - | 4 | 1123 | 30053730 | ||
DV660020.1 | - | 1 | 245 | |||
Y15376.1 | - | 4 | 1119 |