Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025261.1 | NP_001020432.1 | 8 | 1543 | ||
NM_001025262.1 | NP_001020433.1 | 6 | 1474 | |||
NM_001025263.1 | NP_001020434.1 | 7 | 1419 | |||
NM_001025264.1 | NP_001020435.1 | 7 | 1404 | |||
NM_009412.2 | NP_033438.1 | 6 | 1377 | |||
GenBank mRNA | AK005456.1 | - | 6 | 1358 | ||
AK032111.1 | - | 7 | 1382 | |||
AK150935.1 | - | 6 | 1340 | |||
AK151471.1 | - | 6 | 1337 | |||
AK151744.1 | - | 6 | 1337 | |||
AK152543.1 | - | 6 | 1337 | |||
AK153266.1 | - | 6 | 1337 | |||
AK153292.1 | - | 6 | 1348 | |||
AK161753.1 | - | 6 | 1340 | |||
AK168379.1 | - | 6 | 1333 | |||
AK168817.1 | - | 6 | 1341 | |||
AK169751.1 | - | 6 | 1370 | |||
AK170725.1 | - | 6 | 1323 | |||
AK171050.1 | - | 6 | 1318 | |||
AY048852.1 | - | 6 | 1466 | |||
BC004068.1 | - | 6 | 1305 | 3590969 | ||
BC094018.1 | - | 6 | 1327 | 3674995 | ||
U44426.1 | - | 6 | 1306 |