Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165939.1 | NP_001159411.1 | 4 | 330 | ||
NM_001165940.1 | NP_001159412.1 | 3 | 283 | |||
NM_009432.2 | NP_033458.2 | 5 | 371 | |||
GenBank mRNA | BC116829.1 | - | 4 | 292 | 40126148 | |
BC116831.1 | - | 4 | 292 | 40126150 | ||
BC144732.1 | - | 3 | 250 | 9053280 | ||
J00644.1 | - | 4 | 304 | |||
M54943.1 | - | 4 | 304 |