Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009481.2 | NP_033507.2 | 3 | 556 | ||
GenBank mRNA | AK028443.1 | - | 3 | 541 | ||
AK049283.1 | - | 4 | 669 | |||
DQ086491.1 | - | 3 | 517 | |||
U67874.1 | - | 3 | 517 |