Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Was | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009515.2 | NP_033541.1 | 1 | 161 | ||
GenBank mRNA | BY232461.1 | - | 1 | 162 | ||
U29673.1 | - | 1 | 122 | |||
U42471.1 | - | 1 | 122 | |||
U54788.1 | - | 1 | 122 |