Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009513.2 | NP_033539.1 | 3 | 410 | ||
GenBank mRNA | AK138191.1 | - | 3 | 341 | ||
AK153741.1 | - | 2 | 318 | |||
AK160941.1 | - | 3 | 396 | |||
BC052019.1 | - | 2 | 233 | 5708003 | ||
D83206.1 | - | 3 | 363 |