Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285792.1 | NP_001272721.1 | 5 | 1049 | ||
NM_001285794.1 | NP_001272723.1 | 4 | 807 | |||
NM_009519.2 | NP_033545.1 | 3 | 815 | |||
GenBank mRNA | AK031573.1 | - | 3 | 659 | ||
AK077828.1 | - | 4 | 746 | |||
AK138967.1 | - | 4 | 808 | |||
BC125482.1 | - | 3 | 762 | 40129997 | ||
BC125484.1 | - | 3 | 762 | 40129999 | ||
X70800.1 | - | 3 | 822 |