Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009576.2 | NP_033602.2 | 3 | 782 | ||
GenBank mRNA | AK028305.1 | - | 1 | 585 | ||
AK035908.1 | - | 1 | 586 | |||
AK046913.1 | - | 3 | 783 | |||
AK163477.1 | - | 3 | 1856 | |||
D78174.1 | - | 1 | 1035 |