Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018024.2 | NP_001018024.1 | 3 | 729 | ||
GenBank mRNA | BC002600.1 | - | 3 | 691 | 3161408 | |
BT006749.1 | - | 2 | 140 | |||
X64644.1 | - | 3 | 715 |