Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC069735.1 | - | 1 | 326 | 7262624 | |
M62303.1 | - | 1 | 332 | |||
S82362.1 | - | 1 | 1321 |