Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crisp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009638.3 | NP_033768.3 | 5, 6 | 486 | ||
GenBank mRNA | AK009449.1 | - | 5, 6 | 487 | ||
L05559.1 | - | 5, 6 | 456 | |||
M92849.1 | - | 5, 6 | 452 |