Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009633.3 | NP_033763.2 | 1 | 1322 | ||
GenBank mRNA | AF332049.1 | - | 1 | 1028 | ||
AF332050.1 | - | 1 | 1028 | |||
BC066862.1 | - | 1 | 1322 |
Set Membership: |
G-protein coupled receptor |