Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036684.2 | NP_001031761.1 | 2 | 570 | ||
NM_009723.5 | NP_033853.1 | 4 | 759 | |||
GenBank mRNA | AB030737.1 | - | 1 | 131 | ||
AB030738.1 | - | 1 | 131 | |||
AF053471.1 | - | 3 | 588 | |||
AK051287.1 | - | 2 | 565 | |||
AK147240.1 | - | 4 | 645 | |||
AK147357.1 | - | 2 | 499 | |||
AK147369.1 | - | 4 | 695 | |||
AK147399.1 | - | 2 | 734 | |||
AK147501.1 | - | 4 | 706 | |||
BC075643.1 | - | 2 | 570 | 30621322 | ||
GU734816.2 | - | 1 | 131 |