siRNA Details
Gene Symbol: Atrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009730.2 | NP_033860.2 | 2, 3 | 560 | ||
GenBank mRNA | AF116897.1 | - | 2, 3 | 564 | ||
AK155069.1 | - | 2, 3 | 562 | |||
BC141054.1 | - | 2, 3 | 541 | 9055666 |

More Information
Other Species Targeted

Panther Classification:
