Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009766.3 | NP_033896.2 | 1 | 282 | ||
GenBank mRNA | AK038616.1 | - | 1 | 283 | ||
BC106961.1 | - | 1 | 263 | 40039080 | ||
BC106962.1 | - | 1 | 263 | 40039081 |
Set Membership: |
G-protein coupled receptor |