Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009740.2 | NP_033870.1 | 3 | 957 | ||
GenBank mRNA | AF057701.1 | - | 3 | 841 | ||
AF100339.1 | - | 3 | 847 | |||
AF134396.1 | - | 3 | 841 | |||
AJ006289.1 | - | 3 | 838 | |||
AK076082.1 | - | 3 | 869 | |||
AK140179.1 | - | 3 | 829 | |||
AK146236.1 | - | 3 | 862 | |||
AK150883.1 | - | 3 | 858 | |||
AK152563.1 | - | 3 | 858 | |||
AK152847.1 | - | 3 | 854 | |||
AK156890.1 | - | 3 | 911 | |||
AK168084.1 | - | 3 | 862 | |||
AK169126.1 | - | 3 | 864 | |||
AK172158.1 | - | 3 | 868 | |||
BC003703.1 | - | 3 | 876 | |||
BC024379.1 | - | 3 | 835 | 4976147 |