Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009889.2 | NP_034019.1 | 2, 3 | 204 | ||
GenBank mRNA | AF307151.1 | - | 1, 2 | 118 | ||
BC087926.1 | - | 2, 3 | 202 | 30251641 | ||
J00643.1 | - | 2, 3 | 172 |